Gene Transcripts

Gene Symbol
LOC152274
Entrez Gene ID
152274
Gene Name
uncharacterized LOC152274
Gene Aliases
Chromosome Location
Chr. 3: 14799331 - 14811414 on Build GRCh38
UniGene ID
Hs.434128
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_135245.1 3 3365
GenBank mRNA AK056398.1 - 3 3365


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000566553.1 - 3 3365 Chr. 3 - 14799331 - 14811414 [-]

View Full Product Details