Gene Transcripts

Gene Symbol
ZNF205-AS1
Entrez Gene ID
81854
Gene Name
ZNF205 antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 16: 3110460 - 3115598 on Build GRCh38
UniGene ID
Hs.669490
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024166.1 3 1195
NR_024167.1 3 1190
GenBank mRNA BC001809.2 - 3 1195
BC001824.2 - 3 1195

View Full Product Details