Gene Transcripts
- Gene Symbol
- LINC01028
- Entrez Gene ID
- 101928141
- Gene Name
- long intergenic non-protein coding RNA 1028
- Gene Aliases
- TCONS_00025127
- Chromosome Location
- Chr. 17: 70051271 - 70068095 on Build GRCh38
- UniGene ID
- Hs.639899
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_104129.1 |
|
3 |
835 |
|
NR_104130.1 |
|
2 |
713 |
|
GenBank mRNA |
BX647864.1 |
- |
2 |
708 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG185376.1 |
- |
1 |
578 |
|
Chr. 17 - 70067181 - 70067964 [+] |
GENCODE |
ENST00000586373.1 |
- |
2 |
707 |
|
Chr. 17 - 70051277 - 70068095 [+] |
View Full Product Details