Gene Transcripts
- Gene Symbol
- LOC100505851
- Entrez Gene ID
- 100505851
- Gene Name
- uncharacterized LOC100505851
- Gene Aliases
- Chromosome Location
- Chr. 19: 23949579 - 23957930 on Build GRCh38
- UniGene ID
- Hs.190888
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125686.1 |
- |
3 |
1414 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000596326.5 |
- |
3 |
1413 |
|
Chr. 19 - 23919081 - 23957930 [-] |
View Full Product Details