Gene Transcripts
- Gene Symbol
- Gm11691
- Entrez Gene ID
- 100038601
- Gene Name
- predicted gene 11691
- Gene Aliases
- OTTMUSG00000003387
- Chromosome Location
- Chr. 11: 114251843 - 114253672 on Build GRCm38
- UniGene ID
- Mm.308684
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK132953.1 |
- |
1 |
231 |
|
View Full Product Details