Gene Transcripts
- Gene Symbol
- Gm15232
- Entrez Gene ID
- 627268
- Gene Name
- predicted gene 15232
- Gene Aliases
- OTTMUSG00000019548
- Chromosome Location
- Chr. X: 167088093 - 167102110 on Build GRCm38
- UniGene ID
- Mm.391335
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK041611.1 |
- |
4 |
1870 |
|
View Full Product Details