Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_134438.3 | NP_602320.2 | 2 | 908 | |
GenBank mRNA | AB016602.1 | - | 2 | 651 | |
AK158275.1 | - | 2 | 911 | ||
BC098482.1 | - | 2 | 599 | 30103328 | |
BC127619.1 | - | 2 | 532 | 40106370 | |
BC127620.1 | - | 2 | 532 | 40106375 |