Gene Transcripts

Gene Symbol
Mrgprd
Entrez Gene ID
211578
Gene Name
MAS-related GPR, member D
Gene Aliases
Gm499, Mgrd, MrgD, TGR7
Chromosome Location
Chr. 7: 145314835 - 145324059 on Build GRCm38
UniGene ID
Mm.370299
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203490.3 NP_987075.1 2 583
GenBank mRNA AY762266.1 - 2 583
BC137948.1 - 2 489 8860967
BC137950.1 - 2 489 8860969

View Full Product Details