Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013854.1 | NP_038882.1 | 1, 2 | 111 | |
GenBank mRNA | AK077741.1 | - | 1, 2 | 123 | |
BC032250.1 | - | 1, 2 | 104 | ||
BC046965.1 | - | 1, 2 | 85 | 6485365 | |
BC063094.1 | - | 1, 2 | 111 | 6516514 |