Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001134829.1 | NP_001128301.1 | 6, 7 | 1092 | |
NM_172266.3 | NP_758470.1 | 6, 7 | 1071 | ||
GenBank mRNA | AK133814.1 | - | 6, 7 | 1072 | |
AK172914.1 | - | 6, 7 | 1042 | ||
BC013667.1 | - | 6, 7 | 980 | 4020003 | |
BC024750.1 | - | 4, 5 | 562 |