Gene Transcripts

Gene Symbol
Mylk2
Entrez Gene ID
228785
Gene Name
myosin, light polypeptide kinase 2, skeletal muscle
Gene Aliases
9830004H17Rik, BB138278
Chromosome Location
Chr. 2: 152911276 - 152923065 on Build GRCm38
UniGene ID
Mm.250604
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081044.2 NP_001074513.2 11 1923
GenBank mRNA AK079396.1 - 11 1937
BC019408.1 - 6 610
BC158041.1 - 11 1754 9088157

View Full Product Details