Gene Transcripts
- Gene Symbol
- Olfr1281
- Entrez Gene ID
- 257979
- Gene Name
- olfactory receptor 1281
- Gene Aliases
- MOR248-14P, MOR248-18
- Chromosome Location
- Chr. 2: 111328421 - 111329338 on Build GRCm38
- UniGene ID
- Mm.377322
- Species
- Mus musculus
- Gene Symbol
- Olfr1282
- Entrez Gene ID
- 258909
- Gene Name
- olfactory receptor 1282
- Gene Aliases
- MOR248-16, MOR248-2, Olfr1557
- Chromosome Location
- Chr. 2: 111335159 - 111336076 on Build GRCm38
- UniGene ID
- Mm.377746
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146907.2 |
NP_667118.2
|
1 |
680 |
|
- Gene Symbol
- Olfr1286
- Entrez Gene ID
- 277562
- Gene Name
- olfactory receptor 1286
- Gene Aliases
- GA_x5J8B7TU5VU-1563-1372, GA_x6K02T2Q125-72472405-72471488, MOR248-21
- Chromosome Location
- Chr. 2: 111420032 - 111420949 on Build GRCm38
- UniGene ID
- Mm.377872
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207254.1 |
NP_997137.1
|
1 |
680 |
|
View Full Product Details