Gene Transcripts
- Gene Symbol
- Olfr594
- Entrez Gene ID
- 258246
- Gene Name
- olfactory receptor 594
- Gene Aliases
- MOR32-10
- Chromosome Location
- Chr. 7: 103219720 - 103220655 on Build GRCm38
- UniGene ID
- Mm.377390
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207143.1 |
NP_997026.1
|
1 |
809 |
|
View Full Product Details