Gene Transcripts
- Gene Symbol
- PRL
- Entrez Gene ID
- 5617
- Gene Name
- prolactin
- Gene Aliases
- GHA1
- Chromosome Location
- Chr. 6: 22287244 - 22302897 on Build GRCh38
- UniGene ID
- Hs.1905
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000948.5 |
NP_000939.1
|
2, 3 |
724 |
|
NM_001163558.2 |
NP_001157030.1
|
3, 4 |
396 |
|
XM_011514753.2 |
XP_011513055.1
|
3, 4 |
443 |
|
XM_011514754.2 |
XP_011513056.1
|
4, 5 |
456 |
|
GenBank mRNA |
BC015850.1 |
- |
2, 3 |
727 |
|
BC088370.1 |
- |
2, 3 |
254 |
30393428 |
D00411.1 |
- |
1, 2 |
177 |
|
DQ839492.1 |
- |
3, 4 |
112 |
|
GQ305133.1 |
- |
3, 4 |
396 |
|
M29386.1 |
- |
1, 2 |
177 |
|
U75583.1 |
- |
1, 2 |
176 |
|
V00566.1 |
- |
2, 3 |
209 |
|
X54393.1 |
- |
3, 4 |
346 |
|
View Full Product Details