Gene Transcripts
- Gene Symbol
- KLK7
- Entrez Gene ID
- 5650
- Gene Name
- kallikrein related peptidase 7
- Gene Aliases
- PRSS6, SCCE, hK7
- Chromosome Location
- Chr. 19: 50976479 - 50984064 on Build GRCh38
- UniGene ID
- Hs.151254
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001207053.1 |
NP_001193982.1
|
3, 4 |
614 |
|
NM_001243126.1 |
NP_001230055.1
|
3, 4 |
597 |
|
NM_005046.3 |
NP_005037.1
|
4, 5 |
745 |
|
NM_139277.2 |
NP_644806.1
|
4, 5 |
575 |
|
GenBank mRNA |
AF411214.1 |
- |
4, 5 |
719 |
|
AF411215.1 |
- |
3, 4 |
588 |
|
AK289660.1 |
- |
4, 5 |
598 |
|
AK295313.1 |
- |
3, 4 |
597 |
|
AY601109.1 |
- |
3, 4 |
473 |
|
AY646152.1 |
- |
2, 3 |
329 |
|
BC032005.1 |
- |
4, 5 |
584 |
4750452 |
DQ018784.1 |
- |
3, 4 |
473 |
|
KF963189.1 |
- |
3, 4 |
437 |
|
KF963190.1 |
- |
3, 4 |
438 |
|
KF963191.1 |
- |
3, 4 |
438 |
|
KF963193.1 |
- |
3, 4 |
438 |
|
L33404.1 |
- |
3, 4 |
488 |
|
View Full Product Details