Gene Transcripts

Gene Symbol
Nav3
Entrez Gene ID
260315
Gene Name
neuron navigator 3
Gene Aliases
4732483H20Rik, 9630020C08Rik, POMFIL1, Pomfil1p, mKIAA0938, unc53H3
Chromosome Location
Chr. 10: 109681573 - 110000391 on Build GRCm38
UniGene ID
Mm.225050
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081035.1 NP_001074504.1 36 6559
GenBank mRNA AK122404.1 - 25 4369

View Full Product Details