Gene Transcripts

Gene Symbol
Serpina3b
Entrez Gene ID
271047
Gene Name
serine (or cysteine) peptidase inhibitor, clade A, member 3B
Gene Aliases
6A1, A030003A19Rik
Chromosome Location
Chr. 12: 104127531 - 104139546 on Build GRCm38
UniGene ID
Mm.120265
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173024.3 NP_766612.1 2 701
GenBank mRNA AK037160.1 - 2 703
BC125418.1 - 2 684 40129933
BC145808.1 - 2 684 40131111

View Full Product Details