Gene Transcripts

Gene Symbol
Fscb
Entrez Gene ID
623046
Gene Name
fibrous sheath CABYR binding protein
Gene Aliases
EG623046
Chromosome Location
Chr. 12: 64471333 - 64474898 on Build GRCm38
UniGene ID
Mm.189552
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163271.1 NP_001156743.1 1 495
GenBank mRNA EF133693.1 - 1 287

View Full Product Details