Gene Transcripts
- Gene Symbol
- RTN2
- Entrez Gene ID
- 6253
- Gene Name
- reticulon 2
- Gene Aliases
- NSP2, NSPL1, NSPLI, SPG12
- Chromosome Location
- Chr. 19: 45485288 - 45497055 on Build GRCh38
- UniGene ID
- Hs.47517
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005619.4 |
NP_005610.1
|
9, 10 |
1734 |
|
NM_206900.2 |
NP_996783.1
|
8, 9 |
1515 |
|
NM_206901.2 |
NP_996784.1
|
5, 6 |
557 |
|
GenBank mRNA |
AF004222.1 |
- |
9, 10 |
1633 |
|
AF004223.1 |
- |
6, 7 |
783 |
|
AF004224.1 |
- |
5, 6 |
525 |
|
AF038540.1 |
- |
6, 7 |
964 |
|
AK291967.1 |
- |
8, 9 |
1496 |
|
AK314158.1 |
- |
8, 9 |
1498 |
|
BC073874.1 |
- |
9, 10 |
1676 |
5242827 |
View Full Product Details