Gene Transcripts
- Gene Symbol
- Gm9099
- Entrez Gene ID
- 668311
- Gene Name
- predicted gene 9099
- Gene Aliases
- EG668311
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.309
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK080358.1 |
- |
|
944 |
|
View Full Product Details