Gene Transcripts

Gene Symbol
Axin2
Entrez Gene ID
29134
Gene Name
axin 2
Gene Aliases
axil
Chromosome Location
Chr. 10: 97212483 - 97238824 on Build Rnor_6.0
UniGene ID
Rn.162212
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024355.1 NP_077331.1 1 804
GenBank mRNA AF017757.1 - 1 804

View Full Product Details