Gene Transcripts
- Gene Symbol
- Adam7
- Entrez Gene ID
- 29641
- Gene Name
- ADAM metallopeptidase domain 7
- Gene Aliases
- 7, ADAM, ADAM 7, EAP, EAP I, EAPI, I
- Chromosome Location
- Chr. 15: 49418946 - 49467174 on Build Rnor_6.0
- UniGene ID
- Rn.10357
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020301.1 |
NP_064697.1
|
6 |
471 |
|
GenBank mRNA |
X66140.1 |
- |
6 |
471 |
|
View Full Product Details