Gene Transcripts
- Gene Symbol
- Padi1
- Entrez Gene ID
- 54282
- Gene Name
- peptidyl arginine deiminase, type I
- Gene Aliases
- Pdi1
- Chromosome Location
- Chr. 5: 159338850 - 159371313 on Build Rnor_6.0
- UniGene ID
- Rn.30030
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019332.1 |
NP_062205.1
|
2, 3 |
402 |
|
GenBank mRNA |
AB010998.1 |
- |
2, 3 |
402 |
|
View Full Product Details