Gene Transcripts
- Gene Symbol
- Sost
- Entrez Gene ID
- 80722
- Gene Name
- sclerostin
- Gene Aliases
- Chromosome Location
- Chr. 10: 89897087 - 89900131 on Build Rnor_6.0
- UniGene ID
- Rn.95369
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030584.1 |
NP_085073.1
|
1, 2 |
255 |
|
GenBank mRNA |
AF326741.1 |
- |
1, 2 |
255 |
|
View Full Product Details