Gene Transcripts

Gene Symbol
Actr3
Entrez Gene ID
81732
Gene Name
ARP3 actin-related protein 3 homolog (yeast)
Gene Aliases
Arp3
Chromosome Location
Chr. 13: 41695520 - 41738622 on Build Rnor_6.0
UniGene ID
Rn.103326
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031068.1 NP_112330.1 4 351
GenBank mRNA AB292042.1 - 4 275
AB292043.1 - 4 264
BC098014.1 - 4 351 7454624
FQ212256.1 - 4 553

View Full Product Details