Gene Transcripts
- Gene Symbol
- Nell1
- Entrez Gene ID
- 81733
- Gene Name
- NEL-like 1 (chicken)
- Gene Aliases
- Chromosome Location
- Chr. 1: 105349069 - 106218956 on Build Rnor_6.0
- UniGene ID
- Rn.10695
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031069.1 |
NP_112331.1
|
4, 5 |
558 |
|
GenBank mRNA |
U48246.1 |
- |
4, 5 |
558 |
|
View Full Product Details