Gene Transcripts
- Gene Symbol
- Mkln1
- Entrez Gene ID
- 83536
- Gene Name
- muskelin 1, intracellular mediator containing kelch motifs
- Gene Aliases
- Chromosome Location
- Chr. 4: 58693384 - 58817924 on Build Rnor_6.0
- UniGene ID
- Rn.6383
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031359.1 |
NP_112649.1
|
10 |
1096 |
|
GenBank mRNA |
AB046442.1 |
- |
10 |
1096 |
|
View Full Product Details