Gene Transcripts

Gene Symbol
Abcd2
Entrez Gene ID
84356
Gene Name
ATP-binding cassette, subfamily D (ALD), member 2
Gene Aliases
ALDR
Chromosome Location
Chr. 7: 132294564 - 132343169 on Build Rnor_6.0
UniGene ID
Rn.19678
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033352.1 NP_203503.1 1 888
GenBank mRNA AF131294.1 - 1 888

View Full Product Details