Gene Transcripts

Gene Symbol
Gsr
Entrez Gene ID
116686
Gene Name
glutathione reductase
Gene Aliases
Chromosome Location
Chr. 16: 62197617 - 62239987 on Build Rnor_6.0
UniGene ID
Rn.19721
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053906.2 NP_446358.2 4 196
GenBank mRNA FM035154.1 - 4 196
U73174.1 - 323

View Full Product Details