Gene Transcripts

Gene Symbol
TBXA2R
Entrez Gene ID
6915
Gene Name
thromboxane A2 receptor
Gene Aliases
BDPLT13, TXA2-R
Chromosome Location
Chr. 19: 3594506 - 3608749 on Build GRCh38
UniGene ID
Hs.442530
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001060.5 NP_001051.1 3 2457
NM_201636.2 NP_963998.2 4 1797
GenBank mRNA BC121148.1 - 2 1716
BC121149.2 - 920

View Full Product Details