Gene Transcripts
- Gene Symbol
- Lhx5
- Entrez Gene ID
- 124451
- Gene Name
- LIM homeobox 5
- Gene Aliases
- Chromosome Location
- Chr. 12: 41663000 - 41671437 on Build Rnor_6.0
- UniGene ID
- Rn.54653
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_139036.1 |
NP_620605.1
|
2 |
728 |
|
GenBank mRNA |
L35572.1 |
- |
2 |
728 |
|
View Full Product Details