Gene Transcripts
- Gene Symbol
- Samsn1
- Entrez Gene ID
- 170637
- Gene Name
- SAM domain, SH3 domain and nuclear localization signals, 1
- Gene Aliases
- Nash
- Chromosome Location
- Chr. 11: 14254047 - 14304603 on Build Rnor_6.0
- UniGene ID
- Rn.81187
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_130821.1 |
NP_570834.1
|
4, 5 |
410 |
|
GenBank mRNA |
AB067679.1 |
- |
4, 5 |
410 |
|
View Full Product Details