Gene Transcripts

Gene Symbol
TLL1
Entrez Gene ID
7092
Gene Name
tolloid like 1
Gene Aliases
ASD6, TLL
Chromosome Location
Chr. 4: 165873238 - 166105178 on Build GRCh38
UniGene ID
Hs.106513
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204760.1 NP_001191689.1 6 1432
NM_012464.4 NP_036596.3 6 1432
XM_011532214.1 XP_011530516.1 6 1490
XM_017008570.1 XP_016864059.1 7 1558
GenBank mRNA AF282732.1 - 6 1036
BC016922.2 - 6 950
BC136429.1 - 6 1342 9020418
BC136430.1 - 6 1342 9020419
BC144085.1 - 6 1342 9052605
U91963.1 - 6 1432

View Full Product Details