Gene Transcripts

Gene Symbol
Nop14
Entrez Gene ID
289724
Gene Name
NOP14 nucleolar protein
Gene Aliases
Nol14, RGD1305605
Chromosome Location
Chr. 14: 81340141 - 81361277 on Build Rnor_6.0
UniGene ID
Rn.12567
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106013.1 NP_001099483.1 11, 12 1743
XM_006251192.1 XP_006251254.1 11, 12 1748
GenBank mRNA FQ218277.1 - 10, 11 1417

View Full Product Details