Gene Transcripts

Gene Symbol
Mss51
Entrez Gene ID
289904
Gene Name
MSS51 mitochondrial translational activator
Gene Aliases
Zmynd17
Chromosome Location
Chr. 15: 4240203 - 4252072 on Build Rnor_6.0
UniGene ID
Rn.25565
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106025.1 NP_001099495.1 3 382
GenBank mRNA FQ215928.1 - 1 29

View Full Product Details