Gene Transcripts

Gene Symbol
UBE2H
Entrez Gene ID
7328
Gene Name
ubiquitin conjugating enzyme E2 H
Gene Aliases
E2-20K, GID3, UBC8, UBCH, UBCH2
Chromosome Location
Chr. 7: 129830732 - 129952960 on Build GRCh38
UniGene ID
Hs.643548
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001202498.1 NP_001189427.1 3 159
NM_003344.3 NP_003335.1 3 555
NM_182697.2 NP_874356.1 3 555
GenBank mRNA AB451490.1 - 3 150
AK316590.1 - 3 138
AY302138.1 - 3 150
BC006277.2 - 3 467 3957107
BI088146.1 - 3 159
BT006756.1 - 3 150
CB995681.1 - 572
KJ534959.1 - 3 150
Z29328.1 - 3 203
Z29330.1 - 3 203

View Full Product Details