Gene Transcripts

Gene Symbol
Akap13
Entrez Gene ID
293024
Gene Name
A kinase (PRKA) anchor protein 13
Gene Aliases
Chromosome Location
Chr. 1: 137013467 - 137315273 on Build Rnor_6.0
UniGene ID
Rn.2027
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106271.2 NP_001099741.2 6 922
XM_006229382.2 XP_006229444.1 6 1733
XM_006229384.2 XP_006229446.1 6 1733
XM_006229386.2 XP_006229448.1 6 1733

View Full Product Details