Gene Transcripts

Gene Symbol
Uap1l1
Entrez Gene ID
296560
Gene Name
UDP-N-acteylglucosamine pyrophosphorylase 1-like 1
Gene Aliases
Chromosome Location
Chr. 3: 2579250 - 2584523 on Build Rnor_6.0
UniGene ID
Rn.23051
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134516.1 NP_001127988.1 2 454
GenBank mRNA BC168670.1 - 2 454

View Full Product Details