Gene Transcripts

Gene Symbol
LOC300308
Entrez Gene ID
300308
Gene Name
similar to hypothetical protein 4930509O22
Gene Aliases
Chromosome Location
Chr. 7: 23011962 - 23026659 on Build Rnor_6.0
UniGene ID
Rn.68319
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013952.1 NP_001013974.1 1365
GenBank mRNA BC090074.1 - 1365

View Full Product Details