Gene Transcripts

Gene Symbol
Fam214a
Entrez Gene ID
300836
Gene Name
family with sequence similarity 214, member A
Gene Aliases
RGD1310552
Chromosome Location
Chr. 8: 81863619 - 81941548 on Build Rnor_6.0
UniGene ID
Rn.23973
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106838.1 NP_001100308.1 12, 13 3179
XM_008766246.1 XP_008764468.1 11, 12 5186
GenBank mRNA FQ212313.1 - 11, 12 3017

View Full Product Details