Gene Transcripts

Gene Symbol
Lrrc37a
Entrez Gene ID
303556
Gene Name
leucine rich repeat containing 37A
Gene Aliases
Lrrc37a3, RGD1559753
Chromosome Location
Chr. 10: 91548704 - 91581397 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008768302.1 XP_008766524.1 1 1217

View Full Product Details