Gene Transcripts

Gene Symbol
Ildr1
Entrez Gene ID
303914
Gene Name
immunoglobulin-like domain containing receptor 1
Gene Aliases
RGD1307792
Chromosome Location
Chr. 11: 67004036 - 67037184 on Build Rnor_6.0
UniGene ID
Rn.155292
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127536.3 NP_001121008.2 6 833
XM_006248394.2 XP_006248456.1 6 902
GenBank mRNA BC166709.1 - 6 833

View Full Product Details