Gene Transcripts

Gene Symbol
Oraov1
Entrez Gene ID
309136
Gene Name
oral cancer overexpressed 1
Gene Aliases
Chromosome Location
Chr. 1: 218071002 - 218082700 on Build Rnor_6.0
UniGene ID
Rn.15258
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107565.1 NP_001101035.1 2 482
XM_006230782.1 XP_006230844.1 3 289
XM_006230783.1 XP_006230845.1 3 289

View Full Product Details