Gene Transcripts

Gene Symbol
Zfp64
Entrez Gene ID
311661
Gene Name
zinc finger protein 64
Gene Aliases
Chromosome Location
Chr. 3: 165713070 - 165741932 on Build Rnor_6.0
UniGene ID
Rn.6243
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012093.1 NP_001012093.1 6 2022
GenBank mRNA BC085820.1 - 6 2022

View Full Product Details