Gene Transcripts

Gene Symbol
Mcf2
Entrez Gene ID
317598
Gene Name
MCF.2 cell line derived transforming sequence
Gene Aliases
RGD1566098
Chromosome Location
Chr. X: 143159071 - 143292364 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773576.1 XP_008771798.1 5 930

View Full Product Details