Gene Transcripts

Gene Symbol
Naglt1
Entrez Gene ID
337920
Gene Name
Na+ dependent glucose transporter 1
Gene Aliases
Chromosome Location
Chr. 20: 45095232 - 45107532 on Build Rnor_6.0
UniGene ID
Rn.22741
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176080.2 NP_788269.1 4 1479
GenBank mRNA AB089802.1 - 4 1479
BC098651.1 - 4 1466 7130354

View Full Product Details