Gene Transcripts

Gene Symbol
Ngly1
Entrez Gene ID
361014
Gene Name
N-glycanase 1
Gene Aliases
Chromosome Location
Chr. 15: 10405453 - 10455973 on Build Rnor_6.0
UniGene ID
Rn.108552
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014136.1 NP_001014158.1 7 1184
GenBank mRNA BC083837.1 - 7 1184

View Full Product Details