Gene Transcripts

Gene Symbol
Bscl2
Entrez Gene ID
361722
Gene Name
Berardinelli-Seip congenital lipodystrophy 2 (seipin)
Gene Aliases
Chromosome Location
Chr. 1: 225035956 - 225046137 on Build Rnor_6.0
UniGene ID
Rn.105761
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012171.1 NP_001012171.1 4, 5 964
GenBank mRNA BC089942.1 - 4, 5 964
FQ220723.1 - 221

View Full Product Details