Gene Transcripts

Gene Symbol
Oas1h
Entrez Gene ID
494200
Gene Name
2 ' -5 ' oligoadenylate synthetase 1H
Gene Aliases
Chromosome Location
Chr. 12: 41214587 - 41224331 on Build Rnor_6.0
UniGene ID
Rn.162143
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009491.1 NP_001009491.1 4 873
XM_008769247.1 XP_008767469.1 4 950
GenBank mRNA AY196701.1 - 4 873

View Full Product Details