Gene Transcripts

Gene Symbol
Eng
Entrez Gene ID
497010
Gene Name
endoglin
Gene Aliases
Chromosome Location
Chr. 3: 11679530 - 11717485 on Build Rnor_6.0
UniGene ID
Rn.187025
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010968.2 NP_001010968.1 7 1112
GenBank mRNA AY562420.1 - 7 825
BC105860.1 - 7 1112 7456529

View Full Product Details